Skip to Content
Merck
5-Methylfurfural analytical standard

5-Methylfurfural

5-Methylfurfural analytical standard
Synonym(s):
5-Methyl-2-furaldehyde
Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4