![4-Nitrophenyl N,N′-diacetyl-β-D-chitobioside ≥99% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/219/070/ebb722d2-fdb0-4bba-a271-b32f82b458ac/640/ebb722d2-fdb0-4bba-a271-b32f82b458ac.png)
4-Nitrophenyl N,N′-diacetyl-β-D-chitobioside
![4-Nitrophenyl N,N′-diacetyl-β-D-chitobioside ≥99% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/219/070/ebb722d2-fdb0-4bba-a271-b32f82b458ac/640/ebb722d2-fdb0-4bba-a271-b32f82b458ac.png)
Empirical Formula (Hill Notation):
C22H31N3O13
CAS Number:
Molecular Weight:
545.49
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing