4-(N-[2,4-Diamino-6-pteridinylmethyl]amino)benzoic acid sodium salt
Synonym(s):
4-Amino-4-deoxypteroic acid
Linear Formula:
C14H12N7O2Na
CAS Number:
Molecular Weight:
333.28
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing