4-(Methylthio)phenylboronic acid
Synonym(s):
4-(Methylsulfanyl)benzeneboronic acid, 4-(Methylsulfanyl)phenylboronic acid, 4-(Methylthio)benzeneboronic acid
Linear Formula:
CH3SC6H4B(OH)2
CAS Number:
Molecular Weight:
168.02
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing