![4-Methoxy-1-naphthoic acid AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/417/607/1745b7a4-339d-40e1-aef9-c4dda68b6485/640/1745b7a4-339d-40e1-aef9-c4dda68b6485.png)
4-Methoxy-1-naphthoic acid
![4-Methoxy-1-naphthoic acid AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/417/607/1745b7a4-339d-40e1-aef9-c4dda68b6485/640/1745b7a4-339d-40e1-aef9-c4dda68b6485.png)
Synonym(s):
4-Methoxynaphthalene-1-carboxylic acid
Empirical Formula (Hill Notation):
C12H10O3
CAS Number:
Molecular Weight:
202.21
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing