Skip to Content
Merck
4′-Hydroxypropiophenone 98%

4′-Hydroxypropiophenone

4′-Hydroxypropiophenone 98%
Synonym(s):
4′-Hydroxypropiophenone
Linear Formula:
4-(OH)C6H4COC2H5
CAS Number:
Molecular Weight:
150.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing