Skip to Content
Merck
4-Epitetracycline hydrochloride VETRANAL®, analytical standard

4-Epitetracycline hydrochloride

4-Epitetracycline hydrochloride VETRANAL®, analytical standard
Synonym(s):
4-epi-Tetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing