
4-(Diethylamino)but-2-ynyl 2-hydroxy-2,2-diphenylacetate

Synonym(s):
4-(Diethylamino)-2-butyn-1-yl α-hydroxy-α-phenylbenzeneacetate
Empirical Formula (Hill Notation):
C22H25NO3
CAS Number:
Molecular Weight:
351.44
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1