Skip to Content
Merck
(4-Carboxybutyl)triphenylphosphonium bromide 98%

(4-Carboxybutyl)triphenylphosphonium bromide

(4-Carboxybutyl)triphenylphosphonium bromide 98%
Synonym(s):
5-(Triphenylphosphonio)pentanoic acid bromide, Carboxybutyltriphenylphosphonium bromide, NSC 147756, 4-Carboxybutyltriphenylphosphonium bromide
Linear Formula:
(C6H5)3P(CH2)4COOH · Br
CAS Number:
Molecular Weight:
443.31

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing