![4arm-PEG40K-Succinimidyl Succinate average Mn 40,000](/deepweb/assets/sigmaaldrich/product/structures/212/886/828e2f77-5553-4ba5-a58e-699c06f6d37c/640/828e2f77-5553-4ba5-a58e-699c06f6d37c.png)
4arm-PEG40K-Succinimidyl Succinate
![4arm-PEG40K-Succinimidyl Succinate average Mn 40,000](/deepweb/assets/sigmaaldrich/product/structures/212/886/828e2f77-5553-4ba5-a58e-699c06f6d37c/640/828e2f77-5553-4ba5-a58e-699c06f6d37c.png)
Synonym(s):
4arm-PEG-Succinimidyl Succinate
Linear Formula:
C(CH2O(CH2CH2O)nCOCH2CH2COOC4H4O2N)4
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing