![4arm-PEG10K-Succinimidyl Glutarate pentaerythritol core, average Mn 10,000](/deepweb/assets/sigmaaldrich/product/structures/341/765/39e5912d-fe80-4e80-a2ba-d73bc821a9ba/640/39e5912d-fe80-4e80-a2ba-d73bc821a9ba.png)
4arm-PEG10K
![4arm-PEG10K-Succinimidyl Glutarate pentaerythritol core, average Mn 10,000](/deepweb/assets/sigmaaldrich/product/structures/341/765/39e5912d-fe80-4e80-a2ba-d73bc821a9ba/640/39e5912d-fe80-4e80-a2ba-d73bc821a9ba.png)
Synonym(s):
4arm-PEG-Succinimidyl Glutarate
Linear Formula:
C(CH2O(CH2CH2O)nCOCH2CH2CH2COOC4H4O2N)4
CAS Number:
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing