4,4′-Difluorobenzophenone
Synonym(s):
4,4′-Difluorobenzophenone
Linear Formula:
(FC6H4)2CO
CAS Number:
Molecular Weight:
218.20
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing