Skip to Content
Merck
4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 solution 1 μg/mL in acetone, analytical standard

4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 solution

4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 solution 1 μg/mL in acetone, analytical standard
Synonym(s):
3,6,3-Nonylphenol-13C6, 363-NP-13C6, 4-(1-Ethyl-1,4-dimethylpentyl)phenol-13C6 (ring-13C6)
Empirical Formula (Hill Notation):
13C6C9H24O
CAS Number:
Molecular Weight:
226.31
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique