Skip to Content
Merck
3-tert-Butyl-4-hydroxyanisole Pharmaceutical Secondary Standard; Certified Reference Material

3-tert-Butyl-4-hydroxyanisole

3-tert-Butyl-4-hydroxyanisole Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA
Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique