![3-Hydroxyphenylboronic acid ≥95.0%](/deepweb/assets/sigmaaldrich/product/structures/425/581/5f4d93a0-4b9b-4417-8833-a7e5aa2f084a/640/5f4d93a0-4b9b-4417-8833-a7e5aa2f084a.png)
3-Hydroxyphenylboronic acid
![3-Hydroxyphenylboronic acid ≥95.0%](/deepweb/assets/sigmaaldrich/product/structures/425/581/5f4d93a0-4b9b-4417-8833-a7e5aa2f084a/640/5f4d93a0-4b9b-4417-8833-a7e5aa2f084a.png)
Synonym(s):
3-Hydroxybenzeneboronic acid, m-Hydroxybenzeneboronic acid
Linear Formula:
HOC6H4B(OH)2
CAS Number:
Molecular Weight:
137.93
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing