Skip to Content
Merck
3-Hydroxydesloratadine

3-Hydroxydesloratadine

3-Hydroxydesloratadine
Empirical Formula (Hill Notation):
C19H19ClN2O
CAS Number:
Molecular Weight:
326.82

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing