Skip to Content
Merck
3-Chlorobenzoic acid Pharmaceutical Secondary Standard; Certified Reference Material

3-Chlorobenzoic acid

3-Chlorobenzoic acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-Chlorobenzoic acid
Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique