![3-Bromo-2,4,6-trimethylaniline 98%](/deepweb/assets/sigmaaldrich/product/structures/394/934/b1db9eda-6a7a-4594-89c3-94f28f1340ba/640/b1db9eda-6a7a-4594-89c3-94f28f1340ba.png)
3-Bromo-2,4,6-trimethylaniline
![3-Bromo-2,4,6-trimethylaniline 98%](/deepweb/assets/sigmaaldrich/product/structures/394/934/b1db9eda-6a7a-4594-89c3-94f28f1340ba/640/b1db9eda-6a7a-4594-89c3-94f28f1340ba.png)
Linear Formula:
BrC6H(CH3)3NH2
CAS Number:
Molecular Weight:
214.10
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing