3-(Boc-amino)azetidine
Synonym(s):
3-(Boc-amino)azetidine, tert-Butyl 3-azetidinylcarbamate
Empirical Formula (Hill Notation):
C8H16N2O2
CAS Number:
Molecular Weight:
172.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing