3-[Bis(trimethylsilyl)amino]phenylmagnesium chloride solution
Linear Formula:
[(CH3)3Si]2NC6H4MgCl
CAS Number:
Molecular Weight:
296.24
Products
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing