Skip to Content
Merck
3-Aminophenol PESTANAL®, analytical standard

3-Aminophenol

3-Aminophenol PESTANAL®, analytical standard
Synonym(s):
3-Aminophenol
Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing