Skip to Content
Merck
3-(Aminomethyl)oxetane 97%

3-(Aminomethyl)oxetane

3-(Aminomethyl)oxetane 97%
Synonym(s):
3-oxetanemethanamine
Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing