Skip to Content
Merck
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline AldrichCPR

3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline

3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline AldrichCPR
Empirical Formula (Hill Notation):
C11H10F3N3
CAS Number:
Molecular Weight:
241.21

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing