3,4-Di-O-caffeoylquinic acid
Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing