![3,3-Dimethyl-2-butanone for synthesis](/deepweb/assets/sigmaaldrich/product/images/226/150/ae5682b5-0a93-4a66-b99f-825bd46bd774/640/ae5682b5-0a93-4a66-b99f-825bd46bd774.jpg)
3,3-Dimethyl-2-butanone
![3,3-Dimethyl-2-butanone for synthesis](/deepweb/assets/sigmaaldrich/product/images/226/150/ae5682b5-0a93-4a66-b99f-825bd46bd774/640/ae5682b5-0a93-4a66-b99f-825bd46bd774.jpg)
Synonym(s):
3,3-Dimethyl-2-butanone
Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing