Skip to Content
Merck
2-Thiophenepropionic acid 98%

2-Thiophenepropionic acid

2-Thiophenepropionic acid 98%
Synonym(s):
3-(2-Thienyl)propionic acid
Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing