2-Phenylethanol
Synonym(s):
β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA, 2-Phenylethanol
Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing