
2-Octyl-2-dodecenal

Empirical Formula (Hill Notation):
C20H38O
CAS Number:
Molecular Weight:
294.52
Products
Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing