2-Morpholinoacetic acid hydrochloride
Synonym(s):
Morpholin-4-yl-acetic acid hydrochloride
Empirical Formula (Hill Notation):
C6H12ClNO3
CAS Number:
Molecular Weight:
181.62
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing