Skip to Content
Merck
2-Mononitroglycerin solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

2-Mononitroglycerin solution

2-Mononitroglycerin solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C3H7NO5
CAS Number:
Molecular Weight:
137.09

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique