2-Hydroxyethyl salicylate
Synonym(s):
Ethylene glycol monosalicylate, Ethylene glycol salicylate, Salicylic acid 2-hydroxyethyl ester, Salicylic acid ethylene glycol ester
Linear Formula:
HOC6H4COOCH2CH2OH
CAS Number:
Molecular Weight:
182.17
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing