2-Hydroxy-3-methylbenzaldehyde
Synonym(s):
3-Methylsalicylaldehyde, 2,3-Cresotaldehyde, 2-Hydroxy-3-methylbenzaldehyde
Linear Formula:
HOC6H3(CH3)CHO
CAS Number:
Molecular Weight:
136.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing