![2-Fluoro-5-iodopyridine 97%](/deepweb/assets/sigmaaldrich/product/structures/119/464/f9d53115-85c5-4c38-b323-37d41b36b63a/640/f9d53115-85c5-4c38-b323-37d41b36b63a.png)
2-Fluoro-5-iodopyridine
![2-Fluoro-5-iodopyridine 97%](/deepweb/assets/sigmaaldrich/product/structures/119/464/f9d53115-85c5-4c38-b323-37d41b36b63a/640/f9d53115-85c5-4c38-b323-37d41b36b63a.png)
Empirical Formula (Hill Notation):
C5H3FIN
CAS Number:
Molecular Weight:
222.99
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing