
2-(Diethylamino)-ethanol

Synonym(s):
2-(Diethylamino)-ethanol
Linear Formula:
(C2H5)2NCH2CH2OH
CAS Number:
Molecular Weight:
117.19
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing