2-(Bromomethyl)-6-methylpyridine
Synonym(s):
6-Methyl-2-(bromomethyl)pyridine
Empirical Formula (Hill Notation):
C7H8BrN
CAS Number:
Molecular Weight:
186.05
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing