Skip to Content
Merck
2-Bromoacetophenone for GC derivatization, LiChropur™, ≥99.0%

2-Bromoacetophenone

2-Bromoacetophenone for GC derivatization, LiChropur™, ≥99.0%
Synonym(s):
ω-Bromoacetophenone, Phenacyl bromide
Linear Formula:
C6H5COCH2Br
CAS Number:
Molecular Weight:
199.04
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing