
2-Amino-5-methylphenol

Synonym(s):
2-Hydroxy-4-methylaniline, 4-Amino-3-hydroxytoluene, 6-Amino-m-cresol
Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1