Skip to Content
Merck
2-Acetylpyridine ≥99%, FG

2-Acetylpyridine

2-Acetylpyridine ≥99%, FG
Synonym(s):
Methyl 2-pyridyl ketone
Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing