Skip to Content
Merck
2,5-Dimethylphenol ≥99%

2,5-Dimethylphenol

2,5-Dimethylphenol ≥99%
Synonym(s):
2-Hydroxy-p-xylene, p-Xylenol, 2,5-Dimethylphenol
Linear Formula:
2,5-(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing