Skip to Content
Merck
2,4-Dioxopiperidine-1-carboxylic acid tert-butyl ester AldrichCPR

2,4-Dioxopiperidine-1-carboxylic acid tert-butyl ester

2,4-Dioxopiperidine-1-carboxylic acid tert-butyl ester AldrichCPR
Synonym(s):
tert-Butyl 2,4-dioxopiperidine-1-carboxylate
Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing