Skip to Content
Merck
2,4-Dihydroxypyrimidine-5-carboxylic acid 95%

2,4-Dihydroxypyrimidine-5-carboxylic acid

2,4-Dihydroxypyrimidine-5-carboxylic acid 95%
Synonym(s):
Isoorotic acid, Uracil-5-carboxylic acid
Empirical Formula (Hill Notation):
C5H4N2O4
CAS Number:
Molecular Weight:
156.10
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing