Skip to Content
Merck
2,4-Dichlorobenzoic acid PESTANAL®, analytical standard

2,4-Dichlorobenzoic acid

2,4-Dichlorobenzoic acid PESTANAL®, analytical standard
Synonym(s):
2,4-Dichlorobenzoic acid
Linear Formula:
Cl2C6H3CO2H
CAS Number:
Molecular Weight:
191.01
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique