![2,4-Dichloro-5-nitropyrimidine 97%](/deepweb/assets/sigmaaldrich/product/structures/367/287/28078def-9b43-4694-bf05-7a9008197117/640/28078def-9b43-4694-bf05-7a9008197117.png)
2,4-Dichloro-5-nitropyrimidine
![2,4-Dichloro-5-nitropyrimidine 97%](/deepweb/assets/sigmaaldrich/product/structures/367/287/28078def-9b43-4694-bf05-7a9008197117/640/28078def-9b43-4694-bf05-7a9008197117.png)
Empirical Formula (Hill Notation):
C4HCl2N3O2
CAS Number:
Molecular Weight:
193.98
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing