Skip to Content
Merck
2,3-Dimethylhydroquinone 97%

2,3-Dimethylhydroquinone

2,3-Dimethylhydroquinone 97%
Synonym(s):
1,4-Dihydroxy-2,3-dimethylbenzene, 2,3-Dimethyl-1,4-benzenediol
Linear Formula:
(CH3)2C6H2-1,4-(OH)2
CAS Number:
Molecular Weight:
138.16

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing