
2,3,4,6-Tetra-O-benzyl-D-glucopyranose

Empirical Formula (Hill Notation):
C34H36O6
CAS Number:
Molecular Weight:
540.65
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing