2,2′-Bipyrazine
Synonym(s):
2,2′-Bipyrazinyl, 2,2′-Dipyrazinyl
Empirical Formula (Hill Notation):
C8H6N4
CAS Number:
Molecular Weight:
158.16
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing