![2-[[2-[(2-Carboxybenzoyl)amino]ethoxy]methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/981/784/d38231b4-f5c4-4a1b-96a1-4cfe4e8a80b6/640/d38231b4-f5c4-4a1b-96a1-4cfe4e8a80b6.png)
2-[[2-[(2-Carboxybenzoyl)amino]ethoxy]methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester
![2-[[2-[(2-Carboxybenzoyl)amino]ethoxy]methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/981/784/d38231b4-f5c4-4a1b-96a1-4cfe4e8a80b6/640/d38231b4-f5c4-4a1b-96a1-4cfe4e8a80b6.png)
Empirical Formula (Hill Notation):
C28H29ClN2O8
CAS Number:
Molecular Weight:
556.99
Products
Brand
Formula Weight
Manufacturer
Product Line
Application
API Family
Special Grade
Showing 1-1 of 1