![(1S,2R)-(−)-cis-1-Amino-2-indanol 99%](/deepweb/assets/sigmaaldrich/product/structures/384/144/a6b6b757-0f69-4f55-b66d-296d2d06239d/640/a6b6b757-0f69-4f55-b66d-296d2d06239d.png)
(1S,2R)-(−)-cis-1-Amino-2-indanol
![(1S,2R)-(−)-cis-1-Amino-2-indanol 99%](/deepweb/assets/sigmaaldrich/product/structures/384/144/a6b6b757-0f69-4f55-b66d-296d2d06239d/640/a6b6b757-0f69-4f55-b66d-296d2d06239d.png)
Synonym(s):
(1S,2R)-(−)-cis-1-Amino-2-hydroxyindane
Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing