Skip to Content
Merck
1-Aminohydantoin hydrochloride VETRANAL®, analytical standard

1-Aminohydantoin hydrochloride

1-Aminohydantoin hydrochloride VETRANAL®, analytical standard
Empirical Formula (Hill Notation):
C3H5N3O2 · HCl
CAS Number:
Molecular Weight:
151.55

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing