1,2,3,4-Tetra-O-acetyl-α-L-fucopyranose
Synonym(s):
1,2,3,4-Tetra-O-acetyl-α-L(−)-fucose, Tetra-O-acetyl-α-L(−)fucopyranose
Empirical Formula (Hill Notation):
C14H20O9
CAS Number:
Molecular Weight:
332.30
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing